Vanadium(IV) etioporphyrin III oxide

Lieferant: Thermo Scientific

39586.MF
39586.MFEA 436 EUR
39586.MF
Vanadium(IV) etioporphyrin III oxide
Vanadium(IV) etioporphyrin III oxide

CAS No.: 15709-03-2 Molecular Formula: C32H36N4OV Formula Weight: 543.61 Physical Form: Crystalline Appearance: Purple Odour: Odourless MDL No.: MFCD00058901

Formel: C₃₂H₃₆N₄OV
Molecular Weight: 543,61 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00058901
CAS-Nummer: 15709-03-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR