Titan(IV)bromid ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Titantetrabromid

39592.09
39592.09EA 74 EUR
39592.09
Titan(IV)bromid ≥98%
Titan(IV)bromid

Purity: 98% CAS No.: 7789-68-6 Molecular Formula: TiBr4 Formula Weight: 367.54 Physical Form: Crystalline Density: 2.6 Appearance: Orange Melting Point: 39° Boiling Point: 233° Solubility: Soluble in absolute alcohol. Decomposes in water Merck Reference: 14,9477 UN No.: 3260 MDL No.: MFCD00011265

Formel: Br₄Ti
Molecular Weight: 367.5 g/mol
Siedepunkt: 233 °C
Schmelzpunkt: 39
Dichte: 2.6
Lagertemperatur: Raumtemperatur
MDL: MFCD00011265
CAS-Nummer: 7789-68-6
EINECS: 232-185-0
UN: 3260
ADR: 8 II
Merck Index: 12,09617

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR