Tetrachlorbis(tetrahydrofuran)titan(IV) ≥97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Purity: 98%
CAS No.: 31011-57-1
Molecular Formula: C8H16Cl4O2Ti
Formula Weight: 333.91
Flash Point: -17°(1°F)
Physical Form: Powder/crystalline
Sensitivity: Moisture Sensitive
Appearance: Yellow
Melting Point: 126-128°
UN No.: 2925
MDL No.: MFCD00077884
Formel:
C₄H₈Cl₄OTi Schmelzpunkt: 126…128 °C Flammpunkt: –17 °C (1 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00077884 CAS-Nummer: 31011-57-1 UN: 2925 ADR: 4.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...