PAS-Färbekit, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
|
Gefahr
|
395B-1KT
395B-1KTEA
273
EUR
395B-1KT
PAS-Färbekit, Sigma-Aldrich®
PAS (Periodic Acid Schiff) staining kit
Standard procedure and microwave procedure included in package insert.
When treated with periodic acid, glycols are oxidized to aldehydes. After reaction with Schiff′s Reagent (a mixture of pararosaniline and sodium metabisulfite), a pararosaniline adduct
Lagertemperatur:
Kühlschrank |
ADR:
9,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...