1H-Imidazol-1-ylessigsäure 98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 1-Imidazoleacetic acid, Imidazol-1-yl-acetic acid, 1-(Carboxymethyl)imidazole, 2-(1-Imidazolyl)acetic acid

396050010 396050050 396050250
396050010.EA 29.6 EUR
396050010. 396050050. 396050250.
1H-Imidazol-1-ylessigsäure 98%
2-(1H-Imidazol-1-yl)acetic acid
Formel: C₅H₆N₂O₂
Molecular Weight: 126.1133 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01721800
CAS-Nummer: 22884-10-2

Bestellen

Spezifikation Testergebnisse

Appearance white to light yellow powder
Infrared spectrum Conforms
Titration with NaOH ≥97.5 %
Loss on drying ≤2 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR