Zinn(II)phthalocyanin

Lieferant: Thermo Scientific
Gefahr

Synonyme: Tin(ii)phthalocyanine

39607.02
39607.02EA 73.1 EUR
39607.02
Zinn(II)phthalocyanin
Tin(II) phthalocyanine

CAS No.: 15304-57-1 Molecular Formula: C32H16N8Sn Formula Weight: 631.43 Physical Form: Powder UN No.: 3146 MDL No.: MFCD00058867

Formel: C₃₂H₁₆N₈Sn
Molecular Weight: 631,233 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00058867
CAS-Nummer: 15304-57-1
UN: 3146
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR