Tantal(V)tetraethoxid-2,4-pentanedionat ≥99,99% (Metall-Basis)

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Tantalum(V) (tetraethoxy)(acetylacetonate)

39630.04 39630.09 39630.18
39630.04EA 93.4 EUR
39630.04 39630.09 39630.18
Tantal(V)tetraethoxid-2,4-pentanedionat ≥99,99% (Metall-Basis)
Tantal(V)tetraethoxid-2,4-pentanedionat

Purity: 99.99%, CAS No.: 20219-33-4 Molecular Formula: C13H27O6Ta Formula Weight: 460.30 Flash Point: 65°(149°F) Physical Form: Solid Density: 1.5 Sensitivity: Moisture Sensitive Melting Point: 45° Boiling Point: 95°/0.5mm MDL No.: MFCD00145349

Formel: C₁₃H₂₇O₆Ta
Siedepunkt: 95 °C (0,5 mmHg)
Schmelzpunkt: 45 °C
Dichte: 1,5
Flammpunkt: >65 °C (149 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00145349
CAS-Nummer: 20219-33-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR