Tantal(V)tetraethoxid-2,4-pentanedionat ≥99,99% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Tantalum(V) (tetraethoxy)(acetylacetonate)
39630.04
39630.09
39630.18
39630.04EA
93.4
EUR
39630.04
39630.09
39630.18
Tantal(V)tetraethoxid-2,4-pentanedionat ≥99,99% (Metall-Basis)
Tantal(V)tetraethoxid-2,4-pentanedionat
Purity: 99.99%,
CAS No.: 20219-33-4
Molecular Formula: C13H27O6Ta
Formula Weight: 460.30
Flash Point:
Formel:
C₁₃H₂₇O₆Ta Siedepunkt: 95 °C (0,5 mmHg) Schmelzpunkt: 45 °C Dichte: 1,5 Flammpunkt: >65 °C (149 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00145349 CAS-Nummer: 20219-33-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...