Strontiumaluminat ≥99,5% (Metall-Basis)

Lieferant: Thermo Fisher Scientific

39647.14 39647.22
39647.14EA 114 EUR
39647.14 39647.22
Strontiumaluminat ≥99,5% (Metall-Basis)
Strontium aluminate

Purity: 99.5% CAS No.: 12004-37-4 Molecular Formula: SrAl2O4 Formula Weight: 205.58 Physical Form: -100 Mesh Powder Density: 3.559 Appearance: Silver Odour: Odourless MDL No.: MFCD00054054

Formel: Al₂O₄Sr
Molecular Weight: 205.58 g/mol
Dichte: 3.559
Lagertemperatur: Raumtemperatur
MDL: MFCD00054054
CAS-Nummer: 12004-37-4
EINECS: 234-455-3

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR