Lithium isopropoxide 3 M in THF, AcroSeal®

Lieferant: Thermo Scientific
Gefahr

396501000
396501000.EA 101 EUR
396501000.
Lithium isopropoxide 3 M in THF, AcroSeal®
Lithium isopropoxide

Appearance: Clear red Solution

Formel: C₃H₇LiO
Molecular Weight: 66,028 g/mol
Dichte: 0,87 g/cm³
Flammpunkt: –19 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00048286
CAS-Nummer: 2388-10-5
UN: 2924
ADR: 3,II

Bestellen

Spezifikation Testergebnisse

Appearance Clear yellow to red Solution
Strength 2.9 to 3.3 M

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR