Silver tungsten oxide ≥99%
Lieferant: Thermo Fisher Scientific
Synonyme:
Silver tungstate
39661.18
39661.18EA
314
EUR
39661.18
Silver tungsten oxide ≥99%
Silver tungsten oxide
Purity: 99%
CAS No.: 13465-93-5
Molecular Formula: Ag2O4W
Formula Weight: 463.58
Physical Form: Powder
Sensitivity: Light Sensitive
Appearance: Light yellow
Odour: Odourless
Solubility: Insoluble in water. Soluble in potassium cyanide, ammonium hydroxide and nitric acid
MDL No.: MFCD00014147
Formel:
Ag₂O₄W Lagertemperatur: Raumtemperatur |
MDL:
MFCD00014147 CAS-Nummer: 13465-93-5 EINECS: 236-708-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...