Methylamin 33% (w/w) in Ethanol absolut, AcroSeal®

Lieferant: Thermo Scientific
Gefahr

Synonyme: Aminomethan

396731000
396731000.EA 28.2 EUR
396731000.
Methylamin 33% (w/w) in Ethanol absolut, AcroSeal®
Methylamin
Formel: CH₅N
Molecular Weight: 31,06 g/mol
Dichte: 0,756 g/cm³
Flammpunkt: –23 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00008104
CAS-Nummer: 74-89-5
UN: 2924
ADR: 3,II

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless Solution
Infrared spectrum Conforms
Water ≤1.0 %
Titration with HClO4 31.0 to 35.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR