Tantal ≥99,9% (Metall-Basis), Pulver APS ∼2 µm

Lieferant: Thermo Fisher Scientific
Gefahr

39690.06 39690.14 39690.18
39690.06EA 94.8 EUR
39690.06 39690.14 39690.18
Tantal ≥99,9% (Metall-Basis), Pulver APS ∼2 µm
Tantalum

Purity: 99.9% CAS No.: 7440-25-7 Appearance: Black Odour: Odourless UN No.: 3089 MDL No.: MFCD00011252

Formel: Ta
Dichte: 16,6 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011252
CAS-Nummer: 7440-25-7
EINECS: 231-135-5
UN: 3089
ADR: 4.1,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR