1-Propanol, extra trocken 99.5%, AcroSeal®
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
1-Propylalkohol, 1-Hydroxypropan, Ethylcarbinol
396941000
396940010
396941000.EA
37.2
EUR
396941000.
396940010.
1-Propanol, extra trocken 99.5%, AcroSeal®
1-Propanol
Formel:
H₃CCH₂CH₂OH Molecular Weight: 60.1 g/mol Siedepunkt: 97 °C (760 mmHg) Schmelzpunkt: –127 °C Dichte: 0.8 g/cm³ Flammpunkt: 15 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002941 CAS-Nummer: 71-23-8 UN: 1274 ADR: 3 II Merck Index: 13,07934 |
|
Spezifikation Testergebnisse
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Water | ≤0.005 % (Coulometric) |
GC | ≥99.5 % |
Residue after evaporation | ≤0.001 % |
Filtered to 0.2 micrometer | Filtered |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...