Xenondifluorid ≥99,5% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Purity: 99.5%
CAS No.: 13709-36-9
Molecular Formula: XeF2
Formula Weight: 169.30
Physical Form: Crystalline Granules, shipped in teflon
Density: 4.32
Appearance: White
Melting Point: 127-129°
Merck Reference: 14,10072
UN No.: 3087
MDL No.: MFCD00040538
Fluorinating agent
Formel:
F₂Xe Siedepunkt: 114 °C (1013 hPa) Schmelzpunkt: 127…129 °C Dichte: 4,32 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00040538 CAS-Nummer: 13709-36-9 EINECS: 237-251-2 UN: 3087 ADR: 5.1,II Merck Index: 14,10072 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...