Wolfram ≥99,9% (Metall-Basis), Pulver -325 mesh
Lieferant: Thermo Fisher Scientific
|
Achtung
|
39749.22
39749.36
39749.A3
39749.22EA
64.8
EUR
39749.22
39749.36
39749.A3
Wolfram ≥99,9% (Metall-Basis), Pulver -325 mesh
Tungsten
Purity: 99.9%
CAS No.: 7440-33-7
Appearance: Grey
UN No.: 3089
MDL No.: MFCD00011461
Formel:
W Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011461 CAS-Nummer: 7440-33-7 EINECS: 231-143-9 UN: 3089 ADR: 4.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...