Wolfram ≥99,9% (Metall-Basis), Pulver -325 mesh

Lieferant: Thermo Fisher Scientific
Achtung

39749.22 39749.36 39749.A3
39749.22EA 64.8 EUR
39749.22 39749.36 39749.A3
Wolfram ≥99,9% (Metall-Basis), Pulver -325 mesh
Tungsten

Purity: 99.9% CAS No.: 7440-33-7 Appearance: Grey UN No.: 3089 MDL No.: MFCD00011461

Formel: W
Lagertemperatur: Raumtemperatur
MDL: MFCD00011461
CAS-Nummer: 7440-33-7
EINECS: 231-143-9
UN: 3089
ADR: 4.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR