Tin oxide, antimony doped (∼30% Oxide) (w/w) polimerische Anzeichen
Lieferant: Thermo Fisher Scientific
|
Achtung
|
39754.14
39754.14EA
412
EUR
39754.14
Tin oxide, antimony doped (∼30% Oxide) (w/w) polimerische Anzeichen
Tin oxide, antimony doped
Physical Form: Viscous liquid
Density: 1.61
Solubility: Soluble in alcohols
MDL No.: MFCD00011244
Applications: Used in transparent electrical conducting electrodes
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...