Cyclopropylmagnesiumbromid 0.5 M in THF, AcroSeal®

Lieferant: Thermo Scientific
Gefahr

397611000
397611000.EA 270 EUR
397611000.
Cyclopropylmagnesiumbromid 0.5 M in THF, AcroSeal®
Cyclopropylmagnesiumbromid
Formel: C₃H₅BrMg
Molecular Weight: 145,28 g/mol
Flammpunkt: –17 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00672094
CAS-Nummer: 23719-80-4
UN: 3399
ADR: 4.3,I

Bestellen

Spezifikation Testergebnisse

Appearance Colorless to light yellow to dark yellow Solution
Titration Grignard 0.4 to 0.6 M

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR