Zinn(II)chlorid Dihydrat 98-103%, Kristalle, BAKER ANALYZED® ACS, J.T.Baker®
Lieferant: Avantor
|
Gefahr
|
3980-01EA
337.54
EUR
3980-01
3980-05
Zinn(II)chlorid Dihydrat 98-103%, Kristalle, BAKER ANALYZED® ACS, J.T.Baker®
Zinn(II)chlorid Dihydrat
Stannous chloride, dihydrate, crystal suitable for mercury determination.
Formel:
SnCl₂·2H₂O Siedepunkt: 623 °C (1013 hPa) Schmelzpunkt: 246 °C Dichte: 2,71 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149863 CAS-Nummer: 10025-69-1 EINECS: 231-868-0 UN: 3260 ADR: 8,III Merck Index: 13,08861 |
Spezifikation Testergebnisse
Meets ACS Reagent Chemical Requirements | |
For Laboratory,Research,or Manufacturing Use | 98.0 - 103.0% |
Assay (SnCl₂·2H₂O) | 98.0 - 103.0 % |
Solubility in HCl | Passes Test |
Sulfate (SO₄) | Passes Test |
Calcium (Ca) | ≤0.005 % |
Iron (Fe) | ≤0.003 % |
Lead (Pb) | ≤0.01 % |
Magnesium (Mg) | ≤0.01 % |
Potassium (K) | ≤0.005 % |
Sodium (Na) | ≤0.010 % |
Trace Impurities - Arsenic (As) | ≤2.000 ppm |
Trace Impurities - Mercury (Hg) | ≤0.050 ppm |
Meets Reagent Specifications for testing USP/NF monographs |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...