Aluminiumoxid Polierpaste

Lieferant: Thermo Fisher Scientific

Synonyme: Aluminium(III) oxide, Alumina C, Aluminum oxide

39807.LA
39807.LAEA 145 EUR
39807.LA
Aluminiumoxid Polierpaste
Aluminium oxide

CAS No.: 1344-28-1 Molecular Formula: Al2O3 Formula Weight: 101.96 Physical Form: 0.3 Micron Particles Refractive Index: 1.765 Odour: Odourless Boiling Point: 2980° Merck Reference: 14,356 MDL No.: MFCD00003424


Polishing compound for IR crystal windows

Formel: Al₂O₃
Siedepunkt: 2980 °C
Schmelzpunkt: 2045 °C
Dichte: 3,94 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003424
CAS-Nummer: 1344-28-1
EINECS: 215-691-6
REACH: 01-2119529248-35
Merck Index: 14,356

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR