Rhodium(II) octanoate dimer
Lieferant: Thermo Scientific
|
Achtung
|
39825.03
39825.03EA
357
EUR
39825.03
Rhodium(II) octanoate dimer
Rhodium(II) octanoate dimer
CAS No.: 73482-96-9
Molecular Formula: C32H60O8Rh2
Formula Weight: 778.63
Physical Form: Powder
Appearance: Green
Solubility: Soluble in hot alcohol, dichloromethane, toluene and acetic acid, slightly soluble in alcohol
MDL No.: MFCD00064724
Formel:
C₃₂H₆₀O₈Rh₂ Molecular Weight: 778,63 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00064724 CAS-Nummer: 73482-96-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...