3,4-Difluorophenylmagnesium bromide 0.5 M in THF, AcroSeal®
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
399511000
399511000.EA
152
EUR
399511000.
3,4-Difluorophenylmagnesium bromide 0.5 M in THF, AcroSeal®
3,4-Difluorophenylmagnesium bromide
Appearance: Yellow to dark brown Solution
Formel:
C₆H₃BrF₂Mg Molecular Weight: 217.296 g/mol Siedepunkt: 65 °C (760 mmHg) Dichte: 0.965 g/cm³ Flammpunkt: –17 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00672004 CAS-Nummer: 90897-92-0 UN: 2924 ADR: 3 II |
Spezifikation Testergebnisse
Appearance | Yellow to dark brown Solution |
Titration Grignard | 0.40 to 0.60 M |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...