Methanol (15 - < 30%) (30:70) % (v/v), LiChrosolv®, Supelco®

Lieferant: Merck
Gefahr


M-Clarity™ Qualitätsstufe = MQ100

4.80508.9030
4.80508.9030EA 0 EUR
4.80508.9030
Methanol (15 - < 30%) (30:70) % (v/v), LiChrosolv®, Supelco®
Methanol (15 - < 30%)
Formel: CH₄O/H₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD00004595
UN: 1987
ADR: 3,III

Bestellen

Spezifikation Testergebnisse

Density 0.95 g/cm³ (20 °C)
Flash point 35 °C
Storage class 3 Flammable liquids
WGK WGK 1 slightly water endangering
R Phrase R 10-23/24/25-39/23/24/25
S Phrase S 36/37-45


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR