E/E 20% w/w, Emprove® Expert Cleaning biochromatography resins, SAFC®

Lieferant: Merck
Achtung

Synonyme: Ethyl alcohol

Filter 0,2 um

4.80910.9180 4.80910.9010 4.80910.1000 4.80910.9950 4.80910.6180 4.80910.1000_T 4.80910.6180_T 4.80910.9010_T 4.80910.9180_T
4.80910.9180EA 0 EUR
4.80910.9180 4.80910.9010 4.80910.1000 4.80910.9950 4.80910.6180 MERP4.80910.1000_T MERP4.80910.6180_T MERP4.80910.9010_T MERP4.80910.9180_T
E/E 20% w/w, Emprove® Expert Cleaning biochromatography resins, SAFC®
Ethanol
Formel: C₂H₅OH
Molecular Weight: 46.07 g/mol
Siedepunkt: 78.3
Schmelzpunkt: -117
Flammpunkt: 12
Lagertemperatur: Raumtemperatur
MDL: MFCD00003568
CAS-Nummer: 64-17-5
EINECS: 200-578-6
UN: 1170
ADR: 3 III
Merck Index: 12,03806

Bestellen

Spezifikation Testergebnisse

Assay (Ethanol) 19.5 - 20.5 % (w/w)
Total aerobic microbial count (TAMC) ≤ 10 CFU/ml
Total combined yeasts/moulds count (TYMC) ≤ 10 CFU/ml
Bacterial endotoxins ≤ 2.5 I.U./ml
Evaporation residue ≤ 10 mg/l
Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*).
Manufactured with Ethanol absolute (acc. to Ph Eur, BP, JP, USP) & purified water (acc. to Ph Eur).

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR