Wasser, OmniPur® WFI cell culture tested, sterile, Calbiochem®

Lieferant: Merck Millipore Calbiochem

Synonyme: Wasserstoffoxid, Hydrogenoxid

4.86505.1000 4.86505.5000 4.86505.9200 4.86505.9010 4.86505.9020
4.86505.1000EA 26.5 EUR
4.86505.1000 CALB4.86505.5000 4.86505.9200 4.86505.9010 4.86505.9020
Wasser, OmniPur® WFI cell culture tested, sterile, Calbiochem®
Wasser

Suitable for a wide range of applications.


  • WFI quality
  • Sterile purified water
  • Cell culture tested

Formel: H₂O
Molecular Weight: 18.02 g/mol
Siedepunkt: 100
Schmelzpunkt: 0
Lagertemperatur: Raumtemperatur
MDL: MFCD00011332
CAS-Nummer: 7732-18-5
Merck Index: 13,10098

Bestellen

Spezifikation Testergebnisse

Water conductivity (packaged water) Passes Test
Water conductivity (prior to packaging, stage 1) Passes Test
DNase None detected
Oxidisable substances Passes Test
Plating efficiency Passes Test
Protease None detected
RNase None detected
Particulate matter in injections ≤3 Particles/ml ≥25 µm in size
Total Organic Carbon 500 ppb
Sterility Passes Test
Bacterial endotoxin ≤0,025 EU/ml (per master product record)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR