1,1-Dimethylpyrrolidiniumiodid

Lieferant: Merck

4.94203.0500
4.94203.0500EA 0 EUR
4.94203.0500
1,1-Dimethylpyrrolidiniumiodid
1,1-Dimethylpyrrolidinium iodide
Formel: C₆H₁₄IN
CAS-Nummer: 872-44-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR