5-Tetramethylrhodamine-dUTP, 1 mM Solution
Lieferant: Biotium
40001
40001.EA
215
EUR
40001.
5-Tetramethylrhodamine-dUTP, 1 mM Solution
Nukleinsäure-Reagenzien
Nukleotide
5-TAMRA-dUTP (5-Tetramethylrhodamine-dUTP) can be enzymatically incorporated into DNA to make fluorescently labeled probes, and has absorption/emission at 553/577 nm. Note: 5-TAMRA-dUTP may not be compatible with PCR labeling of DNA probes. Supplied as a 1 mM solution in Tris-HCl buffer, pH 7.5.