Triethylphosphat ≥99%

Lieferant: Thermo Fisher Scientific
Achtung

40001.14 40001.22 40001.36
40001.14EA 33.5 EUR
40001.14 40001.22 40001.36
Triethylphosphat ≥99%
Triethylphosphat

Purity: 99+% CAS No.: 78-40-0 Molecular Formula: (C2H5O)3P(O) Formula Weight: 182.16 Flash Point: 115°(240°F) Physical Form: Liquid Density: 1.072 Refractive Index: 1.4050 Odour: Mild Boiling Point: 215° Merck Reference: 14,9674 MDL No.: MFCD00009077

Formel: C₆H₁₅O₄P
Siedepunkt: 215 °C
Schmelzpunkt: –57 °C
Dichte: 1,072
Flammpunkt: 115 °C (240 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00009077
CAS-Nummer: 78-40-0
EINECS: 201-114-5
Merck Index: 14,9674

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR