DMAO 2 mM in DMSO Verwendung in der nuklearen Färbung
Lieferant: Biotium
40012
40012.EA 430 EUR
40012.
DMAO 2 mM in DMSO Verwendung in der nuklearen Färbung
DMAO
DMAO, 2 mM in DMSO is a green-fluorescent nucleic acid dye for staining live bacteria.
  • Green DNA stain for live bacteria
  • Stable for room temperature storage
  • λEx/λEm (with DNA) = 496/528 nm
  • Cell permeability: Membrane permeant
DMAO is a membrane-permeable DNA dye, excellent for staining live bacteria. It is able to stain both gram positive and gram negative strains, and stained cells can be detected by fluorescence microscopy or flow cytometry.
We also offer DMAO paired together with a dead cell stain (Ethidium Homodimer III, or EthD-III) in our Viability/Cytotoxicity Assay Kit for Bacteria Live and Dead Cells (30027).
DMAO staining in live yeast is not nuclear. In live mammalian cells, DMAO shows both nuclear and cytoplasmic staining. We recommend NucSpot® Live or RedDot™1 dyes for nuclear staining of live mammalian cells. See our Cellular Stains Table for more information on how our dyes stain various organisms.
DMAO preferentially binds to dsDNA with green fluorescence (Ex/Em, DNA-bound = 496/528 nm). Unlike the SYBR dyes, DMAO is extremely stable and can be routinely handled and stored at room temperature without degradation.