Kaliumbromat ≥99,8% ACS
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
40013.22
40013.36
40013.22EA
50.3
EUR
40013.22
40013.36
Kaliumbromat ≥99,8% ACS
Potassium bromate
Purity: 99.8% min
CAS No.: 7758-01-2
Molecular Formula: KBrO3
Formula Weight: 167.00
Physical Form: Granular
Density: 3.27
Melting Point: 370° dec.
Solubility: Slightly soluble in cold water. Soluble in boiling water
Merck Reference: 14,7617
UN No.: 1484
MDL No.: MFCD00011359
As an analytical reagent, oxidizing agent
Formel:
KBrO₃ Schmelzpunkt: 370 °C (Zersetzung) Dichte: 3,27 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011359 CAS-Nummer: 7758-01-2 EINECS: 231-829-8 UN: 1484 ADR: 5.1,II Merck Index: 14,7617 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...