Aminoallyl-dUTP (AA-dUTP) sodium salt, lyophilisiertes Pulver

Lieferant: Biotium

40020-1
40020-1EA 321 EUR
40020-1
Aminoallyl-dUTP (AA-dUTP) sodium salt, lyophilisiertes Pulver
Aminoallyl-dUTP (AA-dUTP) sodium salt

5-Aminoallyl-dUTP (AA-dUTP) can be enzymatically incorporated into DNA. The resulting amine-containing DNA can be subsequently labeled with a fluorescent dye, biotin or other haptens via conventional peptide coupling method.


This two-step method for labeling nucleic acids is considerably more economical than the one-step method using a prelabeled dUTP. Available in lyophilized solid form (1 mg), or as a 10 mM solution in TE Buffer (100 uL).


5-Aminoallyl-dUTP (AA-dUTP) in lyophilized form is suitable for long-term storage.

Formel: C₁₂H₁₇N₃Na₃O₁₄P₃
Molecular Weight: 589,17 g/mol
Lagertemperatur: Gefrierschrank
CAS-Nummer: 116840-18-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR