5-Aminoallyl-UTP sodium salt, lyophilisiertes Pulver

Lieferant: Biotium

40021-1
40021-1EA 321 EUR
40021-1
5-Aminoallyl-UTP sodium salt, lyophilisiertes Pulver
5-Aminoallyl-UTP sodium salt

5-Aminoallyl-UTP can be enzymatically incorporated into RNA for subsequent labeling with an amine-reactive fluorescent dye, biotin, or other hapten.


AA-UTP in lyophilized powder form is suitable for long-term storage.


Bestellinformation: Also available as a 10 mM solution in TE Buffer (100 uL).

Formel: C₁₂H₁₇N₃Na₃O₁₅P₃
Lagertemperatur: Gefrierschrank
CAS-Nummer: 112131-73-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR