5-Aminoallyl-UTP sodium salt 10 mM in TE-Puffer
Lieferant: Biotium
Synonyme:
AA-UTP, 5-(3-Aminoallyl)uridine-5'-triphosphate trisodium salt
40021
40021.EA
265
EUR
40021.
5-Aminoallyl-UTP sodium salt 10 mM in TE-Puffer
5-Aminoallyl-UTP sodium salt
5-Aminoallyl-UTP can be enzymatically incorporated into RNA for subsequent labeling with an amine-reactive fluorescent dye, biotin, or other hapten.
Bestellinformation: Also available in lyophilized solid form (1 mg).
Formel:
C₁₂H₁₇N₃Na₃O₁₅P₃ Molecular Weight: 605,17 g/mol Lagertemperatur: Gefrierschrank |
CAS-Nummer:
112131-73-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...