5-Brom-2'-deoxyuridin (Broxuridin) 95% (durch HPLC), weißgrauer Feststoff

Lieferant: Biotium

Synonyme: 5-Bromo-1-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione, 5-Bromo-2'-deoxyuridine, 5-BrdU

40024
40024.EA 103 EUR
40024.
5-Brom-2'-deoxyuridin (Broxuridin) 95% (durch HPLC), weißgrauer Feststoff
5-Brom-2'-deoxyuridin (Broxuridin)

BrdU (5-Bromo-2′-deoxyuridine) can be incorporated into DNA during cell division and subsequently detected by a BrdU antibody. The probe can be used to study cell-cycle kinetics.

Formel: C₉H₁₁BrN₂O₅
Molecular Weight: 307.10 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00006529
CAS-Nummer: 59-14-3
EINECS: 200-415-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR