5-Bromo-2′-deoxyuridine-5′-triphosphate (5-Bromo-dUTP) 10 mM in TE-Puffer

Lieferant: Biotium
Gefahr

40025
40025.EA 173 EUR
40025.
5-Bromo-2′-deoxyuridine-5′-triphosphate (5-Bromo-dUTP) 10 mM in TE-Puffer
5-Bromo-2′-deoxyuridine-5′-triphosphate (5-Bromo-dUTP)

5-Bromo-dUTP (5-Bromo-2′-deoxyuridine-5′-triphosphate) is widely used in TUNEL assay to detect apoptosis cells, and also is a good substrate for reverse transcriptase.

Formel: C₉H₁₁BrN₂Na₃O₁₄P₃
Molecular Weight: 612,99 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00057405
CAS-Nummer: 102212-99-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR