5-Bromouridine-5′-triphosphate sodium salt (BrUTP ) 10 mM in TE-Puffer

Lieferant: Biotium
Achtung

40026
40026.EA 173 EUR
40026.
5-Bromouridine-5′-triphosphate sodium salt (BrUTP ) 10 mM in TE-Puffer
5-Bromouridine-5′-triphosphate (BrUTP )

BrUTP (5-Bromouridine-5′-triphosphate) can be enzymatically incorporated into RNA and detected with anti-BrdU antibodies.

Formel: C₉H₁₁BrN₂Na₃O₁₅P₃
Molecular Weight: 628,98 g/mol
Lagertemperatur: Gefrierschrank
CAS-Nummer: 161848-60-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR