Bor ≥98% (Metall-Basis), kristallines Pulver -325 mesh

Lieferant: Thermo Fisher Scientific
Achtung

40036.09 40036.18
40036.09EA 132 EUR
40036.09 40036.18
Bor ≥98% (Metall-Basis), kristallines Pulver -325 mesh
Boron

Purity: 98% CAS No.: 7440-42-8 Odour: Odourless UN No.: 3178 MDL No.: MFCD00134034

Formel: B
Lagertemperatur: Raumtemperatur
MDL: MFCD00134034
CAS-Nummer: 7440-42-8
EINECS: 231-151-2
UN: 3178
ADR: 4.1,III
Merck Index: 13,01333

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR