Actinomycin D ≥98% (durch HPLC)
Lieferant: Biotium
40038
40038.EA
220
EUR
40038.
Actinomycin D ≥98% (durch HPLC)
Actinomycin D
Actinomycin D is a nonfluorescent GC-selective intercalator.
- Absorption/Emission 240nm & 443nm/none
- Extinction Coefficient >35,000(240nm) & >25,000(443nm)
- Orange red solid soluble in DMSO at 50 mg/mL
- Store at 4°C
Similar to 7-AAD (cat. no. 40038), actinomycin D has also been used for chromosome banding studies.
Formel:
C₆₂H₈₆N₁₂O₁₆ |
MDL:
MFCD00005033 CAS-Nummer: 50-76-0 UN: 3462 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...