3,3'-Diethyloxadicarbocyanine iodide (DODC), high_purity

Lieferant: Biotium
Achtung

Synonyme: 5-(3-Ethylbenzoxazol-2-ylidene)-1-(3-ethylbenzoxazolium-2-yl)-1,3-pentadiene Iodide, DODC Iodide, DiOC2(5), 3-Ethyl-2-[5-(3-ethyl-2-benzoxazolinylidene)-1,3-pentadienyl]benzoxazolium Iodide

40040
40040.EA 119 EUR
40040.
3,3'-Diethyloxadicarbocyanine iodide (DODC), high_purity
3,3'-Diethyloxadicarbocyanine iodide (DODC)

DODC (DiOC2(5)) has been reported to bind to triplex DNA as well as dimeric hairpin quadruplex DNA. Binding of the dye to triplex DNA quenches the fluorescence of the dye and red-shifts the dye absorption peak.


  • λAbs = 582 nm
  • Red solid soluble in DMSO or MeOH
  • Store at 4°C

Formel: C₂₃H₂₃IN₂O₂
Molecular Weight: 486.35 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00011953
CAS-Nummer: 14806-50-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=832c452dd1c16b23f14eaae37baf9a1322dbde39&vid=832c452dd1c16b23f14eaae37baf9a1322dbde39&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR