CF® Dye dCTP conjugates
Lieferant: Biotium
40028
40027
40058
40055
40056
40057
40066
40067
40068
40027-T
40028-T
40055-T
40056-T
40057-T
40058-T
40066-T
40067-T
40068-T
40028.EA
619
EUR
40028.
40027.
40058.
40055.
40056.
40057.
40066.
40067.
40068.
40027-T
40028-T
40055-T
40056-T
40057-T
40058-T
40066-T
40067-T
40068-T
CF® Dye dCTP conjugates
Proteinkonjugate und Markierungen
CF® dye-dCTP can be used to synthesise probes for in situ hybridisation, microarray, or to synthesise labelled DNA probes for in situ hybridisation and nucleic acid blotting applications.
- Synthesise exceptionally bright and photostable fluorescent DNA probes
- Detect by fluorescence microscopy, flow cytometry, or in gel
- Compatible with fixed cells or tissue sections
- Choice of nine CF® dye colours from green to far-red
- CF® dyes have superior brightness, photostability, and are water-soluble
Bitte beachten CF™594 dCTP is not compatible with PCR labelling of DNA probes.