DEAC-dUTP (diethylaminocoumarin-dUTP), lyophilisiertes Pulver
Lieferant: Biotium
40059
40059.EA
535
EUR
40059.
DEAC-dUTP (diethylaminocoumarin-dUTP), lyophilisiertes Pulver
DEAC-dUTP (diethylaminocoumarin-dUTP)
DEAC-dUTP (diethylaminocoumarin-dUTP) can be used to synthesize fluorescently labeled DNA probes for in-situ hybridization, microarray or blotting techniques.
DEAC has absorption at 426 nm and emission at 480 nm.
Bitte beachten Note: The product is not compatible with PCR applications