Stärke, löslich aus Kartoffeln (aus Kartoffel), Pulver, BAKER ANALYZED® ACS, J.T.Baker®
Lieferant: Avantor
Synonyme:
Amylodextrin, Vitex® Iodindikator
4006-01EA
222.39
EUR
4006-01
4006-05
Stärke, löslich aus Kartoffeln (aus Kartoffel), Pulver, BAKER ANALYZED® ACS, J.T.Baker®
Stärke, löslich
For iodometry.
- Lot analysis on label
Formel:
(C₆H₁₀O₅)n Molecular Weight: 162.14 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00082026 CAS-Nummer: 9005-84-9 EINECS: 232-686-4 Merck Index: 13,08878 |
Spezifikation Testergebnisse
Meets ACS Reagent Chemical Requirements | 5.0 - 7.0 |
For Laboratory,Research,or Manufacturing Use | ≤0.4% |
Solubility | Passes Test |
pH of 2% Solution at 25°C | 5.0 - 7.0 |
Residue after Ignition | ≤0.4 % |
Sensitivity | Passes Test |
Meets Reagent Specifications for testing USP/NF monographs |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...