Fluorescein-12-dUTP, lyophilisiertes Pulver
Lieferant: Biotium
40063
40063.EA 535 EUR
40063.
Fluorescein-12-dUTP, lyophilisiertes Pulver
Fluorescein-12-dUTP
Fluorescein-12-dUTP can be used to synthesize fluorecently labeled DNA probes for in-situ hybridization, microarray or blotting techniques.
Fluorescein has absorption/emission at 494/521 nm.
Bitte beachten Store Fluorescein-12-dUTP at –20 °C or –70 °C, protected from light.