Cyanine 555-dUTP, lyophilisierter roter Feststoff
Lieferant: Biotium
Cyanine 555-dUTP can be used to synthesise labeled DNA probes for in-situ hybridization, microarray or blotting techniques.
Red fluorescent Cyanine 555 is structurally identical to Cy®3 and has absorption/emission at 555/565 nm.
Bitte beachten Store Cyanine 555-dUTP at –20 °C or –70 °C, protected from light.
Cy®3 dye is a registered trademark of GE Healthcare.