Succinimidyl ester ethidium bromide DNA/RNA Vergleichsfarbstoff
Lieferant: Biotium
Synonyme:
EB Succinimidyl Ester, Ethidium bromide succinimidyl ester
The amine reactive succinimidyl ester of EB (ethidium bromide) can be conjugated to peptides, proteins, drugs, polymeric materials and biomolecules with primary amine groups. The conjugates are expected to be essentially nonfluorescent until they are able to complex with nucleic acids, resulting in red fluorescence.
- λEx/λEm (with DNA) = 522/593 nm* (EB also has a large absorption peak at 279 nm)
- Red solid soluble in DMSO or DMF
- Store at -20°C desiccated and protect from light
The conjugates might be useful for studies of nucleic acid binding to various biomolecules, such as DNA-binding proteins. It is also possible that conjugates of other biomolecules might be capable of monitoring their transport into the nucleus. EB dye conjugates of solid or semisolid matrices, such as microspheres, magnetic particles or various resins, might be useful for the detection or affinity isolation of nucleic acids.
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...