Succinimidyl ester thiazole orange dye_DNA/RNA binding

Lieferant: Biotium

Synonyme: TO Succinimidyl Ester, Thiazole orange succinimidyl ester

40073
40073.EA 347 EUR
40073.
Succinimidyl ester thiazole orange dye_DNA/RNA binding
Succinimidyl ester thiazole orange

The amine reactive succinimidyl ester of TO (thiazole orange) can be conjugated to peptides, proteins, drugs, polymeric materials and biomolecules with primary amine groups. The conjugates are expected to be essentially nonfluorescent until they are able to complex with nucleic acids, resulting in green fluorescence.


  • Orange solid soluble in DMSO or DMF
  • Absorption / Emission 512nm / 533 nm (with DNA)
  • Correction factor 260nm 0.067
  • Correction factor 280nm 0.136
  • Stored desiccated at -20°C and protect from light, especially when in solution.


The conjugates might be useful for studies of nucleic acid binding to various biomolecules, such as DNA-binding proteins. It is also possible that conjugates of other biomolecules might be capable of monitoring their transport into the nucleus. TO dye conjugates of solid or semisolid matrices, such as microspheres, magnetic particles or various resins, might be useful for the detection or affinity isolation of nucleic acids.

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2dcd64b892900dd5a592435d54035da1cfbc8ce2&vid=34e559701760262875307b1f134384e162478e51&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR