Succinimidyl ester acridine orange DNA/RNA Vergleichsfarbstoff
Lieferant: Biotium
The amine reactive succinimidyl ester of green fluorescent AO (acridine orange) can be conjugated to peptides, proteins, drugs, polymeric materials and biomolecules with primary amine groups. The conjugates then are able to complex with nucleic acids, resulting in green fluorescence nucleic acid conjugate adduct, making them potentially useful for studies of nucleic acid binding to various biomolecules, such as DNA-binding proteins.
- Orange solid soluble in DMSO or DMF
- Absorption / Emission 500nm / 526nm ( with DNA)
- Correction factor 260nm 0.78
- Correction factor 280nm 0.43
- Stored desiccated at -20°C and protect from light, especially when in solution.
It is also possible that conjugates of other biomolecules might be capable of monitoring their transport into the nucleus. AO dye conjugates of solid or semisolid matrices, such as microspheres, magnetic particles or various resins, might be useful for the detection or affinity isolation of nucleic acids.
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...