Digoxigenin-dUTP, alkali stable
Lieferant: Biotium
40078
40078.EA 332 EUR
40078.
Digoxigenin-dUTP, alkali stable
Nukleinsäure-Reagenzien Nukleotide
Digoxigenin-dUTP can be enzymatically incorporated into DNA via nick translation, random priming, or 3'-end terminal labeling to synthesize labeled DNA probes for in-situ hybridization, microarray or blotting techniques. The digoxigenin (DIG) labeled probe could be detected using a labeled anti-digoxigenin antibody.