Diindiumtrisulfat, wasserfrei ≥99,99% (Metall-Basis)

Lieferant: Thermo Fisher Scientific

Synonyme: Diindiumtris(sulfat), Indium(III)sulfat

40102.14 40102.22
40102.14EA 156 EUR
40102.14 40102.22
Diindiumtrisulfat, wasserfrei ≥99,99% (Metall-Basis)
Diindiumtrisulfat

Purity: 99.99% CAS No.: 13464-82-9 Molecular Formula: In2(SO4)3 Formula Weight: 517.83 Physical Form: Solid Density: 3.438 Appearance: Pale grey Odour: Odourless MDL No.: MFCD00011059

Formel: O₁₂In₂S₃
Molecular Weight: 517.83 g/mol
Schmelzpunkt: 585
Dichte: 3.438
Lagertemperatur: Raumtemperatur
MDL: MFCD00011059
CAS-Nummer: 13464-82-9
EINECS: 236-689-1
Merck Index: 13,04978

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR